Documentos clave
EP0030621905
Eppendorf® Lobind Microplate 384/V-PP, 384 wells
white polypropylene, V-bottom black wells, PCR clean, pkg of 80 ea
About This Item
UNSPSC Code:
41122107
NACRES:
NB.15
material:
V-bottom black wells
white polypropylene
size:
384 wells
sterility:
non-sterile
material
V-bottom black wells
white polypropylene
sterility
non-sterile
packaging
pkg of 80 ea
manufacturer/tradename
Eppendorf® 0030621905
technique(s)
PCR: suitable
size
384 wells
General description
Maximize your sample recovery with Eppendorf LoBind Plates. Eppendorf LoBind Plates are designed with a special two-component polymer mix that creates a hydrophilic surface to ensure optimal sample recovery rates. These plates are specifically designed for research or sensitive testing methods where they enable excellent results due to less sample binding to the plate surface.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)