EP0030602307
Eppendorf® Microplate 96/V-PP, 96 wells
white polypropylene, conical bottom clear wells, PCR clean, pkg of 80 ea
About This Item
UNSPSC Code:
41122107
NACRES:
NB.15
material:
conical bottom clear wells
white polypropylene
white polypropylene
size:
96 wells
sterility:
sterile; autoclaved
Productos recomendados
material
conical bottom clear wells
white polypropylene
sterility
sterile; autoclaved
packaging
pkg of 80 ea
manufacturer/tradename
Eppendorf® 0030602307
technique(s)
PCR: suitable
size
96 wells
General description
Pipette, control and recover your samples with ease using Eppendorf Microplates! Designed with the easy-to-read alphanumeric OptiTrack matrix label to identify individual wells, speed up the filling process and significantly reduce the risk of pipetting errors. Eppendorf Microplates are made with high-transparency polypropylene ensuring quick and easy location of samples
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)