Key Documents
EP0030048385
Sigma-Aldrich
Eppendorf® 7 Adapter for Combitips 25ml and 50ml
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
feature
for Combitips 25 mL and 50 mL, Biopur(R)
packaging
pack of 7 ea (individually blister-wrapped)
manufacturer/tradename
0030048385
Eppendorf®
General description
Biopur quality
Application
Liquid handling
Legal Information
Eppendorf is a registered trademark of Eppendorf AG