Merck
C9635
Sigma-Aldrich
DL-p-Chloroamphetamine hydrochloride
≥94% (HPLC)
Sinónimos:
4-Chloro-α-methylphenethylamine hydrochloride, PCA
About This Item
Fórmula lineal:
ClC6H4CH2CH(CH3)NH2·HCl
Número de CAS:
Peso molecular:
206.11
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
assay
≥94% (HPLC)
form
powder
drug control
Home Office Schedule 1; regulated under CDSA - not available from Sigma-Aldrich Canada
color
white to off-white
mp
168-170 °C (lit.)
solubility
DMSO: >20 mg/mL
H2O: >20 mg/mL
storage temp.
−20°C
SMILES string
Cl.CC(N)Cc1ccc(Cl)cc1
InChI
1S/C9H12ClN.ClH/c1-7(11)6-8-2-4-9(10)5-3-8;/h2-5,7H,6,11H2,1H3;1H
InChI key
DZAANUYJOGCNLL-UHFFFAOYSA-N
Biochem/physiol Actions
Neurotoxic on prolonged administration or at high dosage; substrate for the serotonin transporter that releases serotonin from axon terminals by a nonexocytotic mechanism and blocks the reuptake of serotonin; inhibits tryptophan hydroxylase.
pictograms
Skull and crossbonesHealth hazard
signalword
Danger
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Repr. 2 - STOT RE 2 - STOT SE 3
target_organs
Brain, Respiratory system
Storage Class
6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
ppe
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
P Scholze et al.
The Journal of pharmacology and experimental therapeutics, 293(3), 870-878 (2000-06-28)
HEK 293 cells stably expressing the human serotonin transporter (hSERT) were grown on coverslips, preincubated with [(3)H]5-hydroxytryptamine (5-HT), and superfused. Substrates of the hSERT [e.g., p-chloroamphetamine (PCA)], increased the basal efflux of [(3)H]5-HT in a concentration-dependent manner. 5-HT reuptake blockers
Joke Baute et al.
Genome biology, 16, 168-168 (2015-09-12)
To sustain the global requirements for food and renewable resources, unraveling the molecular networks underlying plant growth is becoming pivotal. Although several approaches to identify genes and networks involved in final organ size have been proven successful, our understanding remains
Sander Biesbroek et al.
The American journal of clinical nutrition, 102(1), 146-154 (2015-05-15)
Several methods are used to determine dietary patterns. Hybrid methods incorporate information on nutrient intake or biological factors to extract patterns relevant to disease etiology. We explore differences between patterns derived with 2 hybrid methods with those obtained by a
H Harrington Cleveland et al.
Development and psychopathology, 27(1), 51-67 (2015-02-03)
Data drawn from the in-home subsample of the PROSPER intervention dissemination trial were used to investigate the moderation of intervention effects on underage alcohol use by maternal involvement and candidate genes. The primary gene examined was dopamine receptor D4 (DRD4).
M Réblová et al.
Persoonia, 34, 40-49 (2015-08-05)
Four morphologically similar specimens of an unidentified perithecial ascomycete were collected on decaying wood submerged in fresh water. Phylogenetic analysis of DNA sequences from protein-coding and ribosomal nuclear loci supports the placement of the unidentified fungus together with Achroceratosphaeria in