Documentos clave
Z743301
Julabo® external Pt 100 bath sensors
L × diam. 1200 mm × 6 mm, PTFE-coated stainless steel
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
material
PTFE-coated stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8891006
L × diam.
1200 mm × 6 mm
General description
Pt 100 sensors enable you to reliably measure temperature within Julabo water baths.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)