Z740741
Aldrich® Essentials boiling flask, flat-bottom, ST/NS 24/29
capacity 500 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Productos recomendados
material
borosilicate glass flask
flat bottom
single-neck
packaging
pkg of 1 ea
capacity
500 mL
Categorías relacionadas
General description
Clear borosilicate glass, flat bottomed boiling flasks with top, outer ST ground joint neck. White marking spot and capacity.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)