Z687774
Replacement NMR needle with cone tip for eVol® electronic syringe
for 50 μL, L × O.D. 11.5 cm × 0.5 mm, gauge size 25
About This Item
gauge size
25
manufacturer/tradename
SGE 038060
L × O.D.
11.5 cm × 0.5 mm
Categorías relacionadas
General description
The eVol® NMR electronic syringe facilitates in-tube sample mixing and enables recovery of precious samples allowing them to be stored in alternative vessels (such as inexpensive vials) rather than being left in valuable tubes. The cone tip replacement needles come in multiple length and gauge options to fit your specific needs.
Legal Information
eVol is a registered trademark of SGE Analytical Science Pty Ltd.
For use with
Referencia del producto