Documentos clave
Z648426
Quickfit® bends with vacuum connection
socket joint: ST/NS 14/23, cone joint: ST/NS 14/23
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
cone joint: ST/NS14/23
socket joint: ST/NS14/23
manufacturer/tradename
Quickfit RA 3/11/SC
availability
not available in USA and Canada
not available in USA and Canada
General description
Socket to cone. With screw-thread adapter for flexible tubing.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)