Z552844
Aldrich® vacuum-distilling adapter with SafetyBarb® hose connector
joint: ST/NS 19/22
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Productos recomendados
joint
joint: ST/NS19/22
General description
Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)