Key Documents
Z511250
Sigma-Aldrich
Fritted disc for solid-phase peptide synthesis flask
Coarse
About This Item
UNSPSC Code:
40161507
NACRES:
NB.24
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
diam.
74.5 mm
pore size
40-60 μm