Documentos clave
Z256366
Sigma-Aldrich
Aldrich® bottle jacket
About This Item
UNSPSC Code:
41100000
availability
not available in EU
General description
Provides virtually unbreakable, secondary containment for transport and storage of
chemicals. Meets OSHA Laboratory Standards (29CFR1910-1450).
  • Rugged PE construction
  • Solvent labels and levels clearly visible through jacket
  • Convenient, handle-locking mechanism keeps top and bottom of jacket locked together
  • Fits common 4 L solvent bottles
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
comparable product
Referencia del producto
Descripción
Precios
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)