Documentos clave
TEM-HC300-AU-UL
Sigma-Aldrich
Ultra-Thin Holey Carbon Film Supported Gold Grids
size 300 mesh, box of 25
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25
size
300 mesh
General description
A 4-6 nm ultra-thin holey carbon film supported grids. Gold grids, 300 Mesh, 25/box
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)