Documentos clave
TEM-CF400-AU-TH
Sigma-Aldrich
Carbon Film Supported Gold Square Mesh
Thick, size 400 mesh, box of 50
About This Item
UNSPSC Code:
41111761
packaging
box of 50
size
400 mesh
General description
A 10 nm thick film of pure carbon supported standard squre mesh grids. Gold grid, 400 mesh.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)