Documentos clave
DWK73805-15415
Kimble® Closed Top Linerless Polypropylene Screw Thread Cap
15-415, white screw cap
About This Item
UNSPSC Code:
41100000
NACRES:
NB.17
material
red polypropylene screw cap (Push fit)
white screw cap
packaging
pkg of 1 packs
manufacturer/tradename
Kimble® 73805-15415
fitting
thread
Application
Closure solution for vials and bottles
Features and Benefits
  • Economical, one-piece construction
  • Unique design provides exceptional sealing properties
  • Autoclavable
  • Designed from ASTM Specification E982
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)