Documentos clave
DWK07-1000
Wheaton® Microliter μlplate Shallow Polypropylene
About This Item
UNSPSC Code:
41106200
NACRES:
NB.15
material:
flat bottom
polypropylene plate
round wells
capacity:
350 μL
material
flat bottom
polypropylene plate
round wells
packaging
pkg of 1 ea
manufacturer/tradename
WHEATON® 07-1000
capacity
350 μL
Application
Ideal for sample collection, storage, combinatorial chemistry and library applications
Features and Benefits
  • Premium virgin polypropylene shallow & medium μLplates is your assurance you achieve accurate results, the first time.
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)