BR708124
Discharge tube with recirculation valve for Dispensette® S
Trace Analysis, 10 mL, PT/IR spring (Tantalum Spring), L 90 mm
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
ETFE cap
manufacturer/tradename
BRAND 708124
packaging
1 ea of
L
90 mm
capacity
10 mL
General description
With recirculation valve
Legal Information
Dispensette is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)