Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Duran
®
Petri Dish
Products
Product Category
dishes (5)
Diameter (mm)
Height (mm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
DWK217524801
Duran
®
Petri Dish,
Four-Sectional, H × diam. 100 mm × 20 mm
Expand
View Pricing
DWK217544606
Duran
®
Petri Dish,
Without Sections, H × diam. 100 mm × 15 mm
Expand
View Pricing
DWK217534802
Duran
®
Petri Dish,
Three-Sectional, H × diam. 100 mm × 20 mm
Expand
View Pricing
DWK217504808
Duran
®
Petri Dish,
Half-Sectional, H × diam. 100 mm × 20 mm
Expand
View Pricing
DWK217544803
Duran
®
Petri Dish,
Without Sections, H × diam. 100 mm × 20 mm
Expand
View Pricing