Skip to Content
Merck
All Photos(1)

Key Documents

V800342

Sigma-Aldrich

Potassium chromate

AR, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CrO4
CAS Number:
Molecular Weight:
194.19
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥99.5%
grade:
AR
form:
solid

grade

AR

vapor density

6.7 (vs air)

product line

Vetec

Assay

≥99.5%

form

solid

pH

9-9.8

mp

971 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.01%

cation traces

Ca: ≤0.005%
Na: ≤0.05%

SMILES string

[K+].[K+].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.2K.4O/q;2*+1;;;2*-1

InChI key

XMXNVYPJWBTAHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecylia Wardak
Journal of hazardous materials, 186(2-3), 1131-1135 (2010-12-21)
A new polyvinylchloride membrane sensor for Pb(2+) with solid contact based on ionic liquid has been prepared. The electrode shows a Nernstian response for lead ions over a wide concentration range (1×10(-8) to 1×10(-1) mol L(-1)) and the slope of
Xue Zhou et al.
Toxicology and applied pharmacology, 236(1), 78-84 (2009-04-18)
Occupational exposure to nickel (Ni), chromium (Cr), and arsenic (As) containing compounds has been associated with lung cancer and other adverse health effects. Their carcinogenic properties may be attributable in part, to activation and/or repression of gene expression induced by
M K Purkait et al.
Journal of hazardous materials, 160(2-3), 502-507 (2008-04-18)
Experimental investigations have been carried out to observe the performance of precipitation behavior of potassium permanganate (KMnO4) and potassium dichromate (K2Cr2O7) in cetyl (hexadecyl) pyridineum chloride (CPC) solution. As in the case of Al3+-dodecylbenzenesulfonate systems [P. Somasundaran, K.P. Anathapadmanabhan, M.S.
Ling-na Zeng et al.
Journal of hazardous materials, 246-247, 1-9 (2013-01-02)
Chromium nephrotoxicity (CrNT) is thought to occur through the oxidant lesion mechanism. There is still a lack of specific remedies against CrNT. We primarily screened Chinese herbal medicines with a potential protective effect against CrNT, e.g., rhubarb (Rheum palmatum L.).
Zhihui Yu et al.
Journal of hazardous materials, 167(1-3), 406-412 (2009-02-11)
Experimental measurements have been made on the batch ion exchange of Ca(II) and Mg(II) from potassium chromate solution using cation exchanger of Amberlite IRC 748 as K+ form. The ion exchange behavior of two alkaline-earth metals on the resin, depending

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service