Skip to Content
Merck
All Photos(1)

Key Documents

SAAR5368510

Quinoline

univAR

Synonym(s):

1-Benzazine, 2,3-Benzopyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N
CAS Number:
Molecular Weight:
129.16
Beilstein:
107477
MDL number:
UNSPSC Code:
12352103

vapor density

4.5 (vs air)

vapor pressure

0.07 mmHg ( 20 °C)

product line

univAR

autoignition temp.

896 °F

packaging

glass bottle of 1 × 2.5 L (SAAR5368510LC)

refractive index

n20/D 1.625 (lit.)

bp

113-114 °C/11 mmHg (lit.)
237 °C (lit.)

mp

−17-−13 °C (lit.)

density

1.093 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

c1ccc2ncccc2c1

InChI

1S/C9H7N/c1-2-6-9-8(4-1)5-3-7-10-9/h1-7H

InChI key

SMWDFEZZVXVKRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service