ZRB1826
Anti-B7-H3/CD276 Antibody, clone 2E22 ZooMAb® Rabbit Monoclonal
recombinant, expressed in HEK 293 cells
About This Item
Recommended Products
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2E22, recombinant monoclonal
description
2E22 Clone
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 57.24 kDa
observed mol wt ~90 kDa
purified by
using Protein A
species reactivity
human
species reactivity (predicted by homology)
porcine, monkey
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
isotype
IgG
epitope sequence
N-terminal half
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... CD276(80381)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in U251 cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected B7-H3/CD276 in U251 cell lysate.
Tested applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected B7-H3/CD276 in PC3 cell lysate.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected B7-H3/CD276 in U251 cells.
Affinity Binding Assay: A representative lot of this antibody bound CD276 (B7-H3) with a KD of 7.2 x 10-8 in an affinity binding assay.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected B7-H3/CD276 in human tonsil tissue sections.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service