Z336777
BRAND® microcentrifuge tube, 1.5 mL with lid, PP
transparent, pack of 3,000
Synonym(s):
Micro Tubes with Screw caps, Microcentrifuge Tubes, Screw caps
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
cap (Lid)
polypropylene
sterility
non-sterile
feature
RCF 20,000 × g
packaging
pack of 3,000
manufacturer/tradename
BRAND
H × O.D.
41 mm × 10.75 mm
capacity
1.5 mL
membrane diam. × thickness
8.5 mm × 0.3 mm
color
transparent
Looking for similar products? Visit Product Comparison Guide
General description
Uniform lid membrane thickness ensures trouble-free piercing by the analyser. Lid provides leak-free seal and fits tightly, yet is easily reopened. Matted labeling area with subdivisions for approximate volume determination. Withstands RCF 20000 × g at 20°C for 20 minutes.
* The amber-colored tubes are particularly suitable for light sensitive reagents.
* The amber-colored tubes are particularly suitable for light sensitive reagents.
Other Notes
BR780502 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service