Skip to Content
Merck
All Photos(5)

Key Documents

WH0007589M9

Sigma-Aldrich

Monoclonal Anti-ZNF38 antibody produced in mouse

clone 4B3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DKFZp434L134, Anti-DKFZp686H10254, Anti-NYREN21, Anti-ZSCAN21, Anti-Zipro1, Anti-zinc finger protein 38

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZSCAN21(7589)

General description

The gene ZSCAN21 (zinc finger and SCAN domain containing 21), also referred to as NY-REN-21, encodes a C2H2 type multi-finger protein that contains an N-terminal SCAN (SRE-ZBP, CTfin51, AW-1 (ZNF174), and Number 18 cDNA or ZnF20) domain and a predicted coil at the central region. The SCAN domain has the ability to form homodimers.

Immunogen

ZSCAN21 (AAH47309.1, 1 a.a. ~ 473 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MMTKVLGMAPVLGPRPPQEQVGPLMVKVEEKEEKGKYLPSLEMFRQRFRQFGYHDTPGPREALSQLRVLCCEWLRPEIHTKEQILELLVLEQFLTILPQELQAWVQEHCPESAEEAVTLLEDLERELDEPGHQVSTPPNEQKPVWEKISSSGTAKESPSSMQPQPLETSHKYESWGPLYIQESGEEQEFAQDPRKVRDCRLSTQHEESADEQKGSEAEGLKGDIISVIIANKPEASLERQCVNLENEKGTKPPLQEAGSKKGRESVPTKPTPGERRYICAECGKAFSNSSNLTKHRRTHTGEKPYVCTKCGKAFSHSSNLTLHYRTHLVDRPYDCKCGKAFGQSSDLLKHQRMHTEEAPYQCKDCGKAFSGKGSLIRHYRIHTGEKPYQCNECGKSFSQHAGLSSHQRLHTGEKPYKCKECGKAFNHSSNFNKHHRIHTGEKPYWCHHCGKTFCSKSNLSKHQRVHTGEGEAP

Biochem/physiol Actions

The gene ZSCAN21 (zinc finger and SCAN domain containing 21) encodes a transcription factor that transcriptionally regulates the gene SNCA that encodes α-synuclein in primary neuronal cultures. α-Synuclein is a presynaptic neuronal protein that is associated with Parkinson disease (PD). Therefore, the ZSCAN21 gene may serve as a potential target in the treatment of PD.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectroscopic characterization of the tumor antigen NY-REN-21 and identification of heterodimer formation with SCAND1.
Carneiro FR
Biochemical and Biophysical Research Communications, 343, 260-268 (2006)
Complex Effects of the ZSCAN21 Transcription Factor on Transcriptional Regulation of a-Synuclein in Primary Neuronal Cultures and in Vivo.
Dermentzaki G
The Journal of Biological Chemistry, 291, 8756-8772 (2016)
Functional dissection of the alpha-synuclein promoter: transcriptional regulation by ZSCAN21 and ZNF219.
Clough RL
Journal of Neurochemistry, 110, 1479-1490 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service