Skip to Content
Merck
All Photos(1)

Key Documents

SRP6576

Sigma-Aldrich

CREB Binding Protein (1081-1197) GST tag human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

CBP, CREBBP, cAMP-responsive element-binding protein 1 CREB-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

40.8 kDa (1081-1197 aa + NT GST Tag)

packaging

pkg of 20 and 100 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CREB1(1385)

General description

The acetylation of histone lysine residues plays a crucial role in the epigenetic regulation of gene transcription. A bromodomain is a protein domain that recognizes acetylated lysine residues such as those on the N-terminal tails of histones. This recognition is often a prerequisite for protein-histone association and chromatin remodeling. These domains function in the linking of protein complexes to acetylated nucleosomes, thereby controlling chromatin structure and gene expression. Thus, bromodomains serve as “readers” of histone acetylation marks regulating the transcription of target promoters. The cAMP response element-binding protein (CREB) binding protein (CREBBP) bromodomain has been shown to modulate the stability and function of the tumor suppressor protein p53. CREBBP bromodomain recognizes the acetylated lysine residue 382 on p53. This product contains the bromodomain region of CREBBP.

Physical form

50 mM Tris, pH 7.5, containing 500 mM sodium chloride, 5 mM β-mercaptoethanol and 5% glycerol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service