Skip to Content
Merck
All Photos(1)

Key Documents

SRP6481

Sigma-Aldrich

CTLA4/CD152 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

CELIAC3, GRD4, GSE, ICOS, IDDM12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 14.3 kDa
observed mol wt 25 kDa by SDS-PAGE (DTT-reduced. Ala 37 is the predicted N-terminal.)

packaging

pkg of 20 and 100 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CTLA4(1493)

General description

CTLA4 (cytotoxic T-lymphocyte associated protein 4) or CD152 (cluster of differentiation) is a T cell receptor which belongs to the immunoglobulin (Ig) superfamily or CD28 family. It is expressed on the surface of Th (T helper) cells. This gene is localized to human chromosome 2q33. This protein is composed of a cytosolic domain, a transmembrane region, and a V domain of 116 amino acids. Alternative splicing results in membrane and soluble forms, where membrane forms work as homodimers and soluble isoforms as monomers.

Biochem/physiol Actions

CTLA4 (cytotoxic T-lymphocyte associated protein 4) is responsible for inhibiting T-cell proliferation and facilitates T cell apoptosis. This protein is expressed post-antigen presentation, and thus, is involved in immune and auto-immune disorders. Therefore, it is also implicated in the pathogenesis of various T cell mediated autoimmune disorders. Studies in Baghdadi or Iraqi population show that polymorphism in this gene is linked with susceptibility to diabetes mellitus type I.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The B7-CD28 superfamily.
Sharpe AH and Freeman GJ
Nature Reviews: Immunology, 2(2), 116-126 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service