SRP6262
VEGFR1/Flt-1 human
recombinant, expressed in HEK 293 cells, ≥98% (SDS-PAGE)
Synonym(s):
FLT, VEGFR1
About This Item
Recommended Products
biological source
human
recombinant
expressed in HEK 293 cells
tag
6-His tagged (N-terminus)
Assay
≥98% (SDS-PAGE)
form
lyophilized powder
mol wt
calculated mol wt 50.6 kDa
observed mol wt 55-60 kDa (DTT-reduced. Protein migrates due to glycosylation. Asp 34 is the predicted N-terminus.)
packaging
pkg of 10 μg
impurities
<1 EU/μg endotoxin (LAL test)
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... VEGFR1(2321)
General description
Physical form
Reconstitution
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Read review on VEGF signaling pathway (Vascular endothelial growth factor (VEGF)/VEGF receptor) and find related products.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service