SRP6196
IL-4Rα human
recombinant, expressed in HEK 293 cells, ≥98% (SDS-PAGE)
Synonym(s):
CD124, IL-4 receptor alpha
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
recombinant
expressed in HEK 293 cells
Assay
≥98% (SDS-PAGE)
form
lyophilized powder
mol wt
~24 kDa
packaging
pkg of 10 and 50 μg
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IL4RA(3566)
General description
IL-4 can signal through type I and type II receptor complexes, which share a common gamma chain (γc). The type I receptor contains in addition to the γc an IL-4Rα subunit, where as the type II receptor contains the IL-13Rα. The secreted extracellular domain of IL-4Rα, called sIL-4Rα, binds IL-4 and antagonizes its activity. It plays an important role in regulating the differentiation of naïve CD4+ T cells and class switching to IgG1 and IgE. Recombinant human sIL-4Rα is a 209 amino acid protein which corresponds to the entire extracellular domain of IL-4Rα.
Physical form
Sterile filtered through a 0.2 micron filter. Lyophilized from 1x PBS pH 7.0.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to - 80°C.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service