Skip to Content
Merck
All Photos(1)

Key Documents

SRP3166

Sigma-Aldrich

TACI human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

TNFRSF13B, Transmembrane activator and CAML interactor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

1.0-3.0 μg/mL

mol wt

17.8 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

TACI, a member of the TNF Receptor superfamily, is expressed in the small intestine, spleen, thymus, peripheral blood leukocytes, activated T cells, and resting B cells. TACI binds to both APRIL and BAFF and can stimulate the activation of transcription factors; NF-kappaB, AP-1, and mediates calcineurin dependent activation of NF-AT (nuclear-factor of activated T cells). TACI also plays a key role in the stimulation of B and T cell function. Soluble TACI inhibits APRIL-stimulated proliferation of primary B-cells by blocking the binding of APRIL to the membrane anchored TACI receptor. Recombinant human TACI is a soluble 160 amino acid polypeptide (17.8 kDa) comprising the TNFR homologous cysteine rich extracellular domain of the TACI protein.

Biochem/physiol Actions

TACI, a member of the TNF Receptor superfamily, is expressed in the small intestine, spleen, thymus, peripheral blood leukocytes, activated T cells, and resting B cells. Recombinant human TACI is a soluble 160 amino acid polypeptide (17.8 kDa) comprising the TNFR homologous cysteine rich extracellular domain of the TACI protein.

Sequence

MSGLGRSRRG GRSRVDQEER FPQGLWTGVA MRSCPEEQYW DPLLGTCMSC KTICNHQSQR TCAAFCRSLS CRKEQGKFYD HLLRDCISCA SICGQHPKQC AYFCENKLRS PVNLPPELRR QRSGEVENNS DNSGRYQGLE HRGSEASPAL PGLKLSADQV

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.0 + 50 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabienne Mackay et al.
Current opinion in immunology, 14(6), 783-790 (2002-11-05)
Over two decades of research have increased the interest in factors from the tumor necrosis factor family. The vast majority of these factors are powerful modulators of critical immune functions and participate in pathogenic mechanisms leading to autoimmune disease. This
J A Gross et al.
Nature, 404(6781), 995-999 (2000-05-09)
B cells are important in the development of autoimmune disorders by mechanisms involving dysregulated polyclonal B-cell activation, production of pathogenic antibodies, and co-stimulation of autoreactive T cells. zTNF4 (BLyS, BAFF, TALL-1, THANK) is a member of the tumour necrosis factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service