Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702997

Sigma-Aldrich

Anti-EEA1 Antibody, clone 8S10A7, Rabbit Monoclonal

Synonym(s):

MST105, MSTP105, ZFYVE2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

8S10A7, monoclonal

form

liquid

mol wt

170 kDa

species reactivity

rat, rat, human

concentration

1.40 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MLRRILQRTPGRVGSQGSDLDSSATPINTVDVNNESSSEGFICPQCMKSLGSADELFKHYEAVHDAGNDSGHGGESNLALKRDDVTLLRQEVQDLQASLK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EEA1(8411)

General description

Binds phospholipid vesicles containing phosphatidylinositol 3-phosphate and participates in endosomal trafficking.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human EEA1 (Q15075).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service