Skip to Content
Merck
All Photos(3)

Key Documents

SAB5702722

Sigma-Aldrich

Anti-UAP1 Antibody, clone 5S7M2, Rabbit Monoclonal

Synonym(s):

AGX, AGX1, AGX2, SPAG2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

5S7M2, monoclonal

form

liquid

mol wt

59 kDa

species reactivity

human, rat, rat

concentration

1.35 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

EDEFSPLKNADSQNGKDNPTTARHALMSLHHCWVLNAGGHFIDENGSRLPAIPRSATNGKSETITADVNHNLKDANDVPIQCEISPLISYAGEGLESYVAD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UAP1(6675)

General description

Converts UTP and GlcNAc-1-P into UDP-GlcNAc, and UTP and GalNAc-1-P into UDP-GalNAc. Isoform AGX1 has 2 to 3 times higher activity towards GalNAc-1-P, while isoform AGX2 has 8 times more activity towards GlcNAc-1-P.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 400-500 of human UAP1 (Q16222).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service