Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702695

Sigma-Aldrich

Anti-PPP6C Antibody, clone 6O2N8, Rabbit Monoclonal

Synonym(s):

PPP6C, PP6, PP6C, protein phosphatase 6 catalytic subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

6O2N8, monoclonal

form

liquid

mol wt

34 kDa

species reactivity

rat, human, rat

concentration

0.38 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

AISPRGAGWLFGAKVTNEFVHINNLKLICRAHQLVHEGYKFMFDEKLVTVWSAPNYCYRCGNIASIMVFKDVNTREPKLFRAVPDSERVIPPRTTTPYFL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPP6C(5537)

General description

This gene encodes the catalytic subunit of protein phosphatase, a component of a signaling pathway regulating cell cycle progression. Splice variants encoding different protein isoforms exist. The pseudogene of this gene is located on chromosome X.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 206-305 of human PPP6C (O00743).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service