Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702449

Sigma-Aldrich

Anti-NEDD4 Antibody, clone 7R0M4, Rabbit Monoclonal

Synonym(s):

NEDD4-1, RPF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

7R0M4, monoclonal

form

liquid

mol wt

120 kDa

species reactivity

human

concentration

0.60 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

GDVDVNDWREHTKYKNGYSANHQVIQWFWKAVLMMDSEKRIRLLQFVTGTSRVPMNGFAELYGSNGPQSFTVEQWGTPEKLPRAHTCFNRLDLPPYESFEE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEDD4(4734)

General description

This gene is the founding member of the NEDD4 family of HECT ubiquitin ligases that function in the ubiquitin proteasome system of protein degradation. The encoded protein contains an N-terminal calcium and phospholipid binding C2 domain followed by multiple tryptophan-rich WW domains and, a C-terminal HECT ubiquitin ligase catalytic domain. It plays critical role in the regulation of a number of membrane receptors, endocytic machinery components and the tumor suppressor PTEN. [provided by RefSeq, Jul 2016]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1200-1300 of human NEDD4 (P46934).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service