Skip to Content
Merck
All Photos(8)

Key Documents

SAB4504551

Sigma-Aldrich

Anti-phospho-Synuclein-α (pTyr133) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
ELISA
WB
species reactivity:
mouse, human, rat
technique(s):
ELISA: 1:40000
western blot: 1:500-1:1000
citations:
3

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 14 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr133)

Gene Information

human ... SNCA(6622)

General description

The SNCA (synuclein α) gene codes for a 140-amino acid protein a-synuclein, that is mapped to human chromosome 4q21-23.

Immunogen

The antiserum was produced against synthesized peptide derived from human Synuclein-alpha around the phosphorylation site of Tyr133.

Immunogen Range: 91-140

Biochem/physiol Actions

Synuclein-α is known to induce oligodendrocyte maturation. Accumulation of α-synuclein in oligodendrocytes results in the loss of myelin and causes neurodegeneration, leading to multiple system atrophy. Mutations in the SNCA gene results in Parkinson′s disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

α-Synucleinopathy associated with G51D SNCA mutation: a link between Parkinson's disease and multiple system atrophy
Kiely AP, et al.
Acta Neuropathologica, 125(5), 753-769 (2013)
Genetic etiology of Parkinson disease associated with mutations in the SNCA, PARK2, PINK1, PARK7, and LRRK2 genes: a mutation update
Nuytemans K, et al.
Human Mutation, 31(7), 763-780 (2010)
?-Synuclein-induced myelination deficit defines a novel interventional target for multiple system atrophy.
Ettle B
Acta Neuropathologica, 132(1), 59-75 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service