Skip to Content
Merck
All Photos(4)

Key Documents

SAB2101877

Sigma-Aldrich

Anti-PRDX1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MSP23, Anti-NKEFA, Anti-PAG, Anti-PAGA, Anti-Peroxiredoxin 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB
clone:
polyclonal
species reactivity:
rat, human, rabbit, guinea pig
citations:
1
technique(s):
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

22 kDa

species reactivity

rat, human, rabbit, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRDX1(5052)

Immunogen

Synthetic peptide directed towards the N terminal region of human PRDX1

Biochem/physiol Actions

PRDX1 is a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. It may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene.

Sequence

Synthetic peptide located within the following region: SSGNAKIGHPAPNFKATAVMPDGQFKDISLSDYKGKYVVFFFYPLDFTFV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan T Quintana et al.
The American journal of pathology, 186(8), 1989-2007 (2016-06-21)
The Bcl2-associated anthanogene (BAG) 3 protein is a member of the BAG family of cochaperones, which supports multiple critical cellular processes, including critical structural roles supporting desmin and interactions with heat shock proteins and ubiquitin ligases intimately involved in protein
Laura Gambari et al.
Pharmacological research, 87, 99-112 (2014-07-08)
Hydrogen sulfide (H2S), which recently emerged as a potent regulator of tissues and organs, is broadly produced in mammalian cells but whether it can regulate bone cell function is still elusive. The main objective of this study was to establish
Juan Liu et al.
The Journal of urology, 191(6), 1920-1926 (2014-01-21)
We investigated oxidative damage caused by and antioxidant responses to peroxiredoxins in the mouse testis at different time points after vasectomy. Mice were divided into a sham operated control group and a vasectomized group. Testicular samples were collected 2 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service