Skip to Content
Merck
All Photos(2)

Key Documents

PLA0164

Sigma-Aldrich

Rabbit anti-ATAD5/FRAG1 Antibody, Affinity Purified

Synonym(s):

C17orf41, ELG1, FLJ12735, FRAG1, chromosome 17 open reading frame 41, chromosome fragility associated gene 1, chromosome fragility-associated gene 1 protein, enhanced level of genomic instability 1 homolog, enhanced level of genomic instability 1 homolog (S. cerevisiae)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
species reactivity:
human
application:
IP
PLA
WB
technique(s):
immunoprecipitation (IP): 2- 5 μg/mg
proximity ligation assay: suitable
western blot: 1:2,000- 1:10,000
citations:

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunoprecipitation (IP): 2- 5 μg/mg
proximity ligation assay: suitable
western blot: 1:2,000- 1:10,000

accession no.

NP_079133.3

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

Immunogen

The epitope recognized by PLA0164 maps to a region between residue 775 and 825 of human ATPase family, AAA domain containing 5 using the numbering given in entry NP_079133.3 (GeneID 79915).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

ATAD5/FRAG1 has been identified as a homolog of a subunit of RFC (replication factor C) and has been shown to be down-regulated in response to replicative stress. ATAD5/FRAG1 may play a role in the surveillance of DNA damage and the cell′s decision to survive or activate apoptosis.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service