Skip to Content
Merck
All Photos(1)

Key Documents

GW10801F

Sigma-Aldrich

Anti-RNASE1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-RNS1, Anti-Ribonuclease pancreatic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human
citations:
3
technique(s):
western blot: suitable

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-10801F

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNASE1(6035)

Immunogen

Immunogen Sequence: GI # 4506547, sequence 1-156
Recombinant pancreatic ribonuclease

Application

Anti-RNASE1 antibody produced in chicken is suitable for western blotting at a working dilution of 1:500 and for cell staining at a working dilution of 1:200.

Biochem/physiol Actions

RNASE1 encodes a protein that is a subset of the ribonuclease A superfamily. It is a pancreatic type of secretory ribonuclease. It acts on the 3′-side of pyrimidine bases and cleaves internal phosphodiester RNA bonds. It is a monomer and preferentially acts on ds-RNA over ss-RNA. Human RNAse I is involved in the digestion of dietary RNA, regulation of vascular homeostasis and inactivation of the HIV. It is also involved in the activation of immature dendritic cells and induction of cytokine production.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Tabish Rehman et al.
PloS one, 6(3), e17159-e17159 (2011-03-17)
Human pancreatic ribonuclease (HPR), a member of RNase A superfamily, has a high activity on double stranded (ds) RNA. By virtue of this activity HPR appears to be involved in the host-defense against pathogenic viruses. To delineate the mechanism of
R Jeremy Johnson et al.
Journal of molecular biology, 368(2), 434-449 (2007-03-14)
The ribonuclease inhibitor protein (RI) binds to members of the bovine pancreatic ribonuclease (RNase A) superfamily with an affinity in the femtomolar range. Here, we report on structural and energetic aspects of the interaction between human RI (hRI) and human
K E Kövér et al.
Journal of molecular biology, 379(5), 953-965 (2008-05-23)
Human pancreatic ribonuclease (RNase 1) is expressed in many tissues; has several important enzymatic and biological activities, including efficient cleavage of single-stranded RNA, double-stranded RNA and double-stranded RNA-DNA hybrids, digestion of dietary RNA, regulation of vascular homeostasis, inactivation of the
Victoria J Mar et al.
Pigment cell & melanoma research, 27(6), 1117-1125 (2014-07-22)
Activating mutations in the GTPase RAC1 are a recurrent event in cutaneous melanoma. We investigated the clinical and pathological associations of RAC1(P29S) in a cohort of 814 primary cutaneous melanomas with known BRAF and NRAS mutation status. The RAC1(P29S) mutation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service