Skip to Content
Merck
All Photos(1)

Key Documents

Tufted Deer

96011147, deer lung, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

Tufted Deer, 96011147

biological source

deer lung

packaging

tube of 5 μg 96011147-DNA-5UG
pkg of vial of cells 96011147-1VL

growth mode

Adherent

karyotype

48XY

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Tufted deer lung

Cell Line Description

The fibroblast-like cell line tufted deer has been derived from the lung of a male deer.

Culture Medium

RPMI 1640 + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C Culture attachment appears to be surface dependent, ECACC recommends the use of Corning Plastics.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service