Skip to Content
Merck
All Photos(1)

Key Documents

C9396

Sigma-Aldrich

Z-Gly-Gly-Arg 7-amido-4-methylcoumarin hydrochloride

protease substrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H33N7O7 · HCl
CAS Number:
Molecular Weight:
616.07
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

solubility

methanol: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(C1=C(O2)C=C(NC([C@H](CCCNC(N)=N)NC(CNC(CNC(OCC3=CC=CC=C3)=O)=O)=O)=O)C=C1)=CC2=O.Cl

InChI

1S/C28H33N7O7.ClH/c1-17-12-25(38)42-22-13-19(9-10-20(17)22)34-26(39)21(8-5-11-31-27(29)30)35-24(37)15-32-23(36)14-33-28(40)41-16-18-6-3-2-4-7-18;/h2-4,6-7,9-10,12-13,21H,5,8,11,14-16H2,1H3,(H,32,36)(H,33,40)(H,34,39)(H,35,37)(H4,29,30,31);1H/t21-;/m0./s1

InChI key

AYMJBLVMUDFWTM-BOXHHOBZSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Substrates

Fluorogenic substrate for urokinase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Zimmerman et al.
Proceedings of the National Academy of Sciences of the United States of America, 75(2), 750-753 (1978-02-01)
A direct rate assay for plasminogen activator has been developed using a synthetic fluorogenic peptide substrate, 7-(N-Cbz-glycylglycylargininamido)-4-methylcoumarin trifluoroacetate. The assay correlates well with the standard 125I-labeled fibrin plate assay using highly purified urokinase, culture fluids from WI-38, Chinese hamster vary
Sakthivel Vaiyapuri et al.
PloS one, 5(3), e9687-e9687 (2010-03-20)
Serine proteases are a major component of viper venoms and are thought to disrupt several distinct elements of the blood coagulation system of envenomed victims. A detailed understanding of the functions of these enzymes is important both for acquiring a
Sophie Laine et al.
Molecules (Basel, Switzerland), 26(8) (2021-05-01)
Two DO3A-type ligands conjugated to substrates of urokinase (L3) and caspase-3 (L4) via a propyl-amide linker were synthesized and their lanthanide(III) (Ln3+) complexes studied. A model compound without peptide substrate (L2) and an amine derivative ligand mimicking the state after
Gustavo Lemos Rocha et al.
Plant science : an international journal of experimental plant biology, 288, 110232-110232 (2019-09-16)
Seed coat is the tissue which establishes an interface between the seed inner tissues and external environment. Our group has shown that cowpea seed coat undergoes coordinated events of programmed cell death (PCD) during development. In relation to germinating seed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service