Skip to Content
Merck
All Photos(2)

Key Documents

N3625

Sigma-Aldrich

Naphthol AS-GR phosphate disodium salt

>99% purity (TLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C22H16NO5PNa2
CAS Number:
Molecular Weight:
451.32
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Naphthol AS-GR phosphate disodium salt,

Assay

>99% (TLC)

Quality Level

form

powder

solubility

water: 50 mg/mL, yellow

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

[Na+].[Na+].Cc1ccccc1NC(=O)c2cc3cc4ccccc4cc3cc2OP([O-])([O-])=O

InChI

1S/C22H18NO5P.2Na/c1-14-6-2-5-9-20(14)23-22(24)19-12-17-10-15-7-3-4-8-16(15)11-18(17)13-21(19)28-29(25,26)27;;/h2-13H,1H3,(H,23,24)(H2,25,26,27);;/q;2*+1/p-2

InChI key

VWSQOOQCTMSZBI-UHFFFAOYSA-L

Biochem/physiol Actions

Produces highly insoluble dark green stain in histochemical studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burstone, M.S.
Enz. Histochem., 277-277 (1962)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service