Skip to Content
Merck
All Photos(1)

Key Documents

CRMFRKR

Freezing Point Standard

ASTM D 2386 (-54.7 °C)

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

according to ASTM® 2386

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

jet fuel

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Freezing Point Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM)

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 - 158.0 °F - closed cup

Flash Point(C)

29 - 70 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service