92940
Triphenylantimony(III)
purum, ≥99.0% (HPLC)
Synonym(s):
Antimony(III)triphenyl, Triphenylstibine
About This Item
Recommended Products
grade
purum
Quality Level
Assay
≥99.0% (HPLC)
form
solid
reaction suitability
core: antimony
reagent type: catalyst
bp
377 °C (lit.)
mp
52-54 °C (lit.)
53-55 °C
SMILES string
c1ccc(cc1)[Sb](c2ccccc2)c3ccccc3
InChI
1S/3C6H5.Sb/c3*1-2-4-6-5-3-1;/h3*1-5H;
InChI key
HVYVMSPIJIWUNA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service