Skip to Content
Merck
All Photos(1)

Key Documents

60470

Sigma-Aldrich

Potassium permanganate

purum p.a., ≥99.0% (RT), fine crystals

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KMnO4
CAS Number:
Molecular Weight:
158.03
EC Number:
MDL number:
UNSPSC Code:
12352312
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (RT)

grade

purum p.a.

Assay

≥99.0% (RT)

form

fine crystals

reaction suitability

reagent type: oxidant

anion traces

chloride, chlorate (as Cl-): ≤50 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Ca: ≤200 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤100 mg/kg
Na: ≤300 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[K+].[O-][Mn](=O)(=O)=O

InChI

1S/K.Mn.4O/q+1;;;;;-1

InChI key

VZJVWSHVAAUDKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for heterogeneous oxidations.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Repr. 2 - Skin Corr. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 1097-1097 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 227-227 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 1047-1047 (1994)
Crown polyether chemistry. Potassium permanganate oxidations in benzene.
Sam DJ and Simmons HE.
Journal of the American Chemical Society, 94(11), 4024-4025 (1972)
Eagleson M.
Concise Encyclopedia Chemistry, 622-622 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service