Skip to Content
Merck
All Photos(1)

Key Documents

45793

Supelco

9-Methylanthracene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12
CAS Number:
Molecular Weight:
192.26
Beilstein:
1907463
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

196-197 °C/12 mmHg (lit.)

mp

77-79 °C (lit.)

density

1.066 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1c2ccccc2cc3ccccc13

InChI

1S/C15H12/c1-11-14-8-4-2-6-12(14)10-13-7-3-5-9-15(11)13/h2-10H,1H3

InChI key

CPGPAVAKSZHMBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

K Takegoshi et al.
Solid state nuclear magnetic resonance, 11(3-4), 189-196 (1998-08-07)
The locus of the photodimerization reaction of 9-methylanthracene in the crystal was examined by high-resolution solid-state 13C NMR techniques. Examination of the 13C spectra of the products showed that only the trans dimer is formed by the solid state photodimerization
H S Lamparczyk et al.
Xenobiotica; the fate of foreign compounds in biological systems, 16(4), 325-333 (1986-04-01)
The metabolism of 9-methylanthracene by liver microsomal preparations from 3-methylcholanthrene-treated rats was examined. The principal metabolites identified were 9-hydroxymethylanthracene, the 1,2- and 3,4-dihydrodiols of the parent hydrocarbon and the 3,4-dihydrodiol of the 9-hydroxymethyl derivative. A small amount of a product
Samantha L Jenkins et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 62(4-5), 1131-1139 (2005-06-15)
Reaction of single crystals of benzoic and trans-cinnamic acids with 200 Torr pressure of ammonia gas in a sealed glass bulb at 20 degrees C generates the corresponding ammonium salts; there is no sign of any 1:2 adduct as has
Klaus Pittertschatscher et al.
Analytical biochemistry, 308(2), 300-306 (2002-11-07)
A novel one-step ethylchloroformate (ECF) derivatization of histamine in biological liquid matrices that allows the sensitive quantification by gas chromatography and mass spectroscopic detection (GC-MS) from small volumes of blood plasma or cell culture supernatants within 15 min is described.
Kamlesh Awasthi et al.
The journal of physical chemistry. A, 113(40), 10603-10609 (2009-10-02)
Photoluminescence of electron donor-acceptor pairs that show photoinduced electron transfer (PIET) has been measured in a polymer film under simultaneous application of electric field and magnetic field. Fluorescence emitted from the locally excited state (LE fluorescence) of 9-methylanthracene (MAnt) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service