OP01
Anti-v-H-Ras (Ab-1) Rat mAb (Y13-259)
liquid, clone Y13-259, Calbiochem®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
biological source
rat
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
Y13-259, monoclonal
form
liquid
contains
≤0.1% sodium azide as preservative
species reactivity
rat, human, mouse
manufacturer/tradename
Calbiochem®
storage condition
do not freeze
isotype
IgG1
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
General description
Purified rat monoclonal antibody generated by immunizing Sprague Dawlay rats with the specified immunogen and fusing splenocytes with Y3Ag1.2.3 rat myeloma cells. Recognizes the ~21 kDa Ras protein (see comments).
Recognizes both phosphorylated and nonphosphorylated forms of v-H-Ras, and v-K-Ras in SW480 and Y1 cells and normal skin tissue. Also recognizes H, K, and N-Ras human translated products.
This Anti-v-H-Ras (Ab-1) Rat mAb (Y13-259) is validated for use in Frozen Sections, Immunoblotting, IF, IP, Paraffin Sections, Neutralization Studies for the detection of v-H-Ras (Ab-1).
Immunogen
Epitope: within amino acids 62-76
recombinant p21 Ras
Application
Frozen Sections (5 µg/ml)
Immunoblotting (2.5 µg/ml, colormetric or chemiluminescence)
Immunofluorescence (2.5 µg/ml)
Immunoprecipitation (use Cat. No. OP01A or 1 µg antibody per sample)
Paraffin Sections (5 µg/ml, saponin or pepsin pre-treatment required)
Neutralization Studies (see application references)
Immunoblotting (2.5 µg/ml, colormetric or chemiluminescence)
Immunofluorescence (2.5 µg/ml)
Immunoprecipitation (use Cat. No. OP01A or 1 µg antibody per sample)
Paraffin Sections (5 µg/ml, saponin or pepsin pre-treatment required)
Neutralization Studies (see application references)
Packaging
Please refer to vial label for lot-specific concentration.
Warning
Toxicity: Standard Handling (A)
Physical form
In 0.05 M sodium phosphate buffer, 0.2% gelatin.
Analysis Note
Positive Control
SW480 or Y1 cells or normal skin tissue
SW480 or Y1 cells or normal skin tissue
Other Notes
Broadly reactive with the phosphorylated and non-phosphorylated forms of v-H-Ras and v-K-Ras (see application references) and recognizes the p21 translational products of the H-, K-, and N-ras human genes (see application references). This antibody does not cross-react with rap. Neutralizes the biological and biochemical activities of H-, K-, and N-ras p21 of vertebrates by binding to residues Glu-63, Ser-65, Ala-66, Met-67, Gln-70, and Arg-73 (see application references). 15 µl v-H-ras (Ab-1) agarose conjugate (Cat. No. OP01A) is recommended to be used as an immunoconcentrator prior to immunoblot analysis of p21ras with another antibody. To immunoprecipitate with the unconjugated antibody, use Protein A with a rabbit anti-rat IgG or Protein G PLUS with a goat anti-rat IgG. Protein A alone will not immunoprecipitate rat IgG. A doublet may be detected corresponding to ras protein with and without farnesyl group. Antibody should be titrated for optimal results in individual systems.
Shimizu, K., et al. 1983. Proc. Natl. Acad. Sci. USA80, 2112.
Taparowsky, E., et al. 1983. Cell34, 581.
Ellis, R.W., et al. 1981. Nature292, 506.
Shih, T.Y., et al. 1980. Nature287, 686.
Taparowsky, E., et al. 1983. Cell34, 581.
Ellis, R.W., et al. 1981. Nature292, 506.
Shih, T.Y., et al. 1980. Nature287, 686.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service