MABX190
Anti- Insulin-like growth factor binding protein 4 (IGFBP-4) clone 190
clone 190, from mouse
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
190, monoclonal
species reactivity
human
technique(s)
ELISA: suitable
western blot: suitable
isotype
IgG1
UniProt accession no.
Gene Information
human ... IGFBP4(3487)
General description
Antibody recognizes the Human insulin like growth factor binding protein 4 (IGFBP-4). IGFBP-4 is a member of the Insulin like growth factor family and is responsible for binding insulin like growth factors I and II. Research has linked levels of C-term (CT-IGFBP-4) and N-term (NT-IGFBP-4) to certain cardiac events, offering potential as a novel cardiac biomarker
Specificity
Human
Application
Confirmed matched pairing. Capture Antibody MABX4180-1KC; Detector Antibody MABX190-1KC
Detect IGFBP-4 using this mouse monoclonal Anti- Insulin-like growth factor binding protein 4 (IGFBP-4) clone 190 Antibody, Cat. No. MABX190-1KC, validated for use in WB and ELISA .
Research Category
Signaling
Signaling
Physical form
Format: Purified
Protein A purified
Purified mouse monoclonal in buffer containing PBS (pH 7.4) with 0.09% sodium azide.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service