Skip to Content
Merck
All Photos(2)

Key Documents

ABS1675

Sigma-Aldrich

Anti-PXDN/VPO1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Peroxidasin homolog, hsPxd01, Melanoma-associated antigen MG50, p53-responsive gene 2 protein, Vascular peroxidase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

bovine, mouse, human

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Peroxidasin homolog (EC 1.11.1.7; UniProt Q92626; also known as hsPxd01, Melanoma-associated antigen MG50, p53-responsive gene 2 protein, Vascular peroxidase 1) is encoded by the PXDN (also known as COPOA, KIAA0230, MG50, PRG2, VPO, VPO1) gene (Gene ID 7837) in human. Human peroxidasin 1 (hsPxd01) belong to the superfamily of heme peroxidases with important roles in innate immunity and hormone biosynthesis, members of which also include myeloperoxidase (MPO), eosinophil peroxidase (EPO), lactoperoxidase (LPO), and thyroid peroxidase (TPO). Peroxidase 1 is a multidomain heme peroxidase that uses bromide as a cofactor in catalyzing the formation of sulfilimine bonds in collagen IV via release of hypohalous acids. Peroxidase 1 plays an essential role in tissue development and architecture by catalyzing sulfilimine bonds formation that confers critical structural reinforcement to collagen IV scaffolds. Human peroxidasin 1 is produced with a signal peptide sequence (a.a. 1-26), the remvoal of which yields the mature protein. In its N-terminal region, hsPxd01 contains four leucine-rich (LRR) repeats (a.a.87-180) flanked by two cysteine-rich domains, the LRRNT (a.a. 27-63) and LRRCT (a.a. 192-245), this is followed by four C2-type Ig-like domains, and a C-terminal von Willebrand factor type C domain (VWFC; a.a. 1413-1471) found in many secreted extracellular matrix proteins.

Specificity

This polyclonal antibody was raised against an N-terminal region sequence present in both spliced isoforms of human VPO1/PXDN protein (UniProt Q92626), but not conserved in MPO, LPO, EPO and TPO (Cheng, G., et al. (2011). Free Radic. Biol. Med. 51(7):1445-1453).

Immunogen

Epitope: LRRNT domain.
KLH-conjugated linear peptide corresponding to a LRRNT domain sequence of human PXDN/VPO1.

Application

Immunohistochemistry Analysis: A 1:400 diluttion from a representative lot detected VPO1 in mouse lung tissue sections (Courtesy of Guangjie Cheng, M.D., Emory University, GA, U.S.A.).
Immunoprecipitation Analysis: A representative lot immunoprecipitated all the radiolabelled VPO1 from the culture supernatant, but not from cell lysates, 20 hrs after an initial 20-min [32S]-Met pulse labeling of HEK293 cells stably expressing transfected VPO1 (Cheng, G., et al. (2011). Free Radic. Biol. Med. 51(7):1445-1453).
Western Blotting Analysis: A representative lots detected cellular VPO1 in total cell lysate and secreted VPO1 in concentrated culture medium following histone deacetylase inhibition by NaBu (Cat. No. 567430) treatment of HEK293 cells stably expressing exogenously transfected VPO1 (Cheng, G., et al. (2011). Free Radic. Biol. Med. 51(7):1445-1453).
Western Blotting Analysis: A representative lot detected VPO1 in human, mouse, and bovine sera samples, as well as a time-dependent cellular VPO1 upregulation in lysates from LPS- or TNF-alpha-stimulated HUVECs (Cheng, G., et al. (2011). Free Radic. Biol. Med. 51(7):1445-1453).
This Anti-PXDN Antibody is validated for use in Western Blotting, Immunohistochemistry (Paraffin), Immunoprecipitation for the detection of PXDN.

Quality

Evaluated by Western Blotting Analysis in human plasma.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected VPO1 in 10 µg of human plasma.

Target description

>165 kDa observed. 162.4/165.3 kDa (human isoform 1 mature/pro-form), 77.50/80.33 kDa (human isoform 2 mature/pro-form), 162.4/165.1 kDa (mouse mature/pro-form) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Purified rabbit polyclonal antibody in 0.1 M Tris-glycine buffer containing 0.02% sodium azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Cao et al.
Redox biology, 45, 102031-102031 (2021-06-12)
Reactive oxygen species (ROS) derived from NADPH oxidases (NOX) plays an essential role in advanced glycation end products (AGEs)-induced diabetic vascular endothelial dysfunction. Peroxidasin (PXDN, VPO1) is one member of peroxidases family that catalyzes hydrogen peroxide (H2O2) to hypochlorous acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service