Skip to Content
Merck
All Photos(1)

Key Documents

14-665

Sigma-Aldrich

MSSK1 Protein, active, 10 µg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405

description

protein target MSSK1

Quality Level

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... SRPK3(26576)

General description

FUNCTION: SwissProt: Q9UPE1 # Required for normal muscle development (By similarity).
SIZE: 533 amino acids; 58997 Da
TISSUE SPECIFICITY: Exclusively expressed in skeletal and heart muscle.
SIMILARITY: SwissProt: Q9UPE1 ## Belongs to the protein kinase superfamily. Ser/Thr protein kinase family. & Contains 1 protein kinase domain.

Quality

routinely evaluated by phosphorylation of PKCtide (#12-356).

Physical properties

65.7kDa

Storage and Stability

6 months at -70°C from date of shipment

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service