Skip to Content
Merck
All Photos(1)

Key Documents

07-546

Sigma-Aldrich

Anti-ADAP/SLAP-130/Fyb Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IP
WB
species reactivity:
mouse
technique(s):
immunoprecipitation (IP): suitable
western blot: suitable
citations:
8

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FYB(2533)

Specificity

ADAP/SLAP-130/Fyb

Immunogen

synthetic peptide corresponding to amino acids 755-768 of mouse Fyn binding protein (ADAP/SLAP-130/Fyb)

Application

Anti-ADAP/SLAP-130/Fyb Antibody is an antibody against ADAP/SLAP-130/Fyb for use in WB & IP.
Research Category
Signaling
Research Sub Category
Immunological Signaling

Kinases & Phosphatases

Quality

routinely evaluated by immunoblot in RIPA lysates from RAW 264.7 macrophage cells

Target description

130kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein A purified

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Signaling by Fyn-ADAP via the Carma1-Bcl-10-MAP3K7 signalosome exclusively regulates inflammatory cytokine production in NK cells.
Rajasekaran, K; Kumar, P; Schuldt, KM; Peterson, EJ; Vanhaesebroeck, B; Dixit, V; Thakar et al.
Nature Immunology null
A J Hunter et al.
Journal of immunology (Baltimore, Md. : 1950), 164(3), 1143-1147 (2000-01-21)
The role of integrin-mediated signaling events in T cell function remains incompletely characterized. We report here that alpha4beta1 integrin stimulation of H9 T cells and normal human T cell blasts results in rapid and transient tyrosine phosphorylation of the adapter
L Geng et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(20), 11527-11532 (2001-09-13)
Aggregation of the high-affinity IgE receptor (FcepsilonRI) on mast cells activates a tyrosine phosphorylation cascade that is required for adhesion and degranulation events leading to the release of histamine and other inflammatory mediators. The full range of intracellular mediators that
N J Boerth et al.
The Journal of biological chemistry, 275(7), 5143-5152 (2000-02-15)
T cell antigen receptor (TCR) engagement results in protein-tyrosine kinase activation which initiates signaling cascades leading to induction of the interleukin-2 gene. Previous studies identified two substrates of the TCR-induced protein-tyrosine kinases, SH2 domain-containing leukocyte specific protein of 76 kDa
Chunyang Li et al.
EMBO molecular medicine, 7(6), 754-769 (2015-04-09)
PD-1 negatively regulates CD8(+) cytotoxic T lymphocytes (CTL) cytotoxicity and anti-tumor immunity. However, it is not fully understood how PD-1 expression on CD8(+) CTL is regulated during anti-tumor immunotherapy. In this study, we have identified that the ADAP-SKAP55 signaling module

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service