07-2184
Anti-acetyl-GAPDH (Lys160) Antibody
0.5 mg/mL, from rabbit
Synonym(s):
Glyceraldehyde-3-phosphate dehydrogenase, GAPDH, Peptidyl-cysteine S-nitrosylase GAPDH
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
species reactivity (predicted by homology)
canine (based on 100% sequence homology), horse (based on 100% sequence homology), bovine (based on 100% sequence homology), sheep (based on 100% sequence homology), feline (based on 100% sequence homology), porcine (based on 100% sequence homology), rat (based on 100% sequence homology), nonhuman primates (based on 100% sequence homology), mouse (based on 100% sequence homology)
concentration
0.5 mg/mL
technique(s)
immunoprecipitation (IP): suitable
inhibition assay: suitable (peptide)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
acetylation (Lys160)
Gene Information
human ... GAPDH(2597)
General description
Immunogen
Application
Immunoprecipitation Analyis: A representative lot immunoprecipitated acetyl GAPDH (Lys60) from LPS/IFNgamma treated RAW264.7 cell lysate (Prof. Akira Sawa, Johns Hopkins University, Baltimore.).
Metabolism
Insulin/Energy Signaling
Quality
Western Blotting Analysis: 1 µg/mL of this antibody detected acetyl GAPDH (Lys60) in LPS/IFNgamma treated RAW264.7 cell lysate and not in untreated RAW264.7 cell lysate.
Target description
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service