TEM-FCF400NI50
Formvar/Carbon Supported Nickel Grids
size 400 mesh, box of 50 ×
Synonym(s):
Formvar/Carbon 400 Mesh, Nckeli, 50pk
All Photos(1)
About This Item
UNSPSC Code:
41111761
packaging
box of 50 ×
size
400 mesh
General description
A formvar coated TEM grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Thickness: 10nm Formvar and 1nm Carbon
Thickness: 10nm Formvar and 1nm Carbon
Choose from one of the most recent versions:
Certificates of Analysis (COA)