DWKWGW535VTML
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting
label, Ethanol (in English, French, Spanish), capacity 250 mL
Synonym(s):
WGW535VTML
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103209
NACRES:
NB.43
feature
label Ethanol (in English, French, Spanish)
packaging
pkg of 5 ea
manufacturer/tradename
Azlon WGW535VTML
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Featuring patented Driplok vapour venting technology, AZLON® multi-lingual safety venting wash bottles ensure the safe and accurate dispensing of liquids. Available in 11 languages and with 6 solvent labels, you can be sure to find the perfect wash bottle for you.
Application
Ideal for ethanol dispensing within the laboratory
Features and Benefits
Patented Driplok vapour venting valve to prevent solvent drips. Multilingual with solvent labels including all new globally harmonised symbols, NFPA diamond, risk phrases and safety information
Legal Information
AZLON is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service